Assay Details
| Gene Symbol: AT2G16730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127225.3 | NP_179264.2 | 11 - 12 | 1542 | 120 | |
| GenBank mRNA | AJ270309.1 | - | 11 - 12 | 1363 | 120 | |
More Information
Set Membership: |
|

AT2G16730
Genomic Map