Assay Details
| Gene Symbol: UBC23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335502.1 | NP_001324377.1 | 6 - 7 | 2960 | 116 | |
| NM_127245.3 | NP_179284.1 | 6 - 7 | 2949 | 116 | ||
More Information
Set Membership: |
|

UBC23
Genomic Map