Assay Details
Gene Symbol: Kin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127276.3 | NP_565408.1 | 5 - 6 | 1245 | 67 | |
NM_201749.1 | NP_973478.1 | 5 - 6 | 1219 | 67 | ||
GenBank mRNA | AF370585.1 | - | 5 - 6 | 814 | 67 | |
AY049263.1 | - | 5 - 6 | 1213 | 67 | ||
AY149966.1 | - | 5 - 6 | 814 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
