Assay Details
| Gene Symbol: MLO7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127298.5 | NP_179335.3 | 6 - 7 | 790 | 150 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MLO7| Gene Symbol: MLO7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127298.5 | NP_179335.3 | 6 - 7 | 790 | 150 | |
Set Membership: |
|