Assay Details
Gene Symbol: MLO8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127302.3 | NP_565416.1 | 4 - 5 | 1047 | 123 | |
GenBank mRNA | AF369569.1 | - | 4 - 5 | 662 | 123 | |
BT002918.1 | - | 4 - 5 | 942 | 123 | ||
BT004356.1 | - | 4 - 5 | 620 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
