Assay Details
Gene Symbol: MLO8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127302.3 | NP_565416.1 | 5 - 6 | 1100 | 104 | |
GenBank mRNA | AF369569.1 | - | 5 - 6 | 715 | 104 | |
BT002918.1 | - | 5 - 6 | 995 | 104 | ||
BT004356.1 | - | 5 - 6 | 673 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
