Assay Details
| Gene Symbol: ATSERAT3;1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127318.5 | NP_565421.1 | 7 - 8 | 1057 | 81 | |
| GenBank mRNA | AF112303.1 | - | 7 - 8 | 811 | 81 | |
| AK176573.1 | - | 7 - 8 | 804 | 81 | ||
| BT024717.1 | - | 7 - 8 | 788 | 81 | ||
More Information
Set Membership: |
|

ATSERAT3;1
Genomic Map