Assay Details
| Gene Symbol: NIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127329.2 | NP_179366.1 | 6 - 7 | 822 | 106 | |
| GenBank mRNA | DQ446513.1 | - | 6 - 7 | 822 | 106 | |
More Information
Set Membership: |
|

NIP1
Genomic Map