Assay Details
Gene Symbol: VPS35A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127333.5 | NP_179370.2 | 12 - 13 | 1532 | 112 | |
GenBank mRNA | BT008553.1 | - | 12 - 13 | 1410 | 112 | |
BT008664.1 | - | 12 - 13 | 1460 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
