Assay Details
Gene Symbol: TAF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202618.2 | NP_001189547.1 | 2 - 3 | 236 | 86 | |
NM_127356.1 | NP_179391.1 | 2 - 3 | 177 | 86 | ||
GenBank mRNA | AY463616.1 | - | 2 - 3 | 177 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
