Assay Details
| Gene Symbol: PIN1AT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127360.4 | NP_179395.1 | 1 - 2 | 640 | 90 | |
| GenBank mRNA | AF360318.1 | - | 1 - 2 | 375 | 90 | |
| AY056314.1 | - | 1 - 2 | 234 | 90 | ||
| AY057514.1 | - | 1 - 2 | 361 | 90 | ||
| AY085059.1 | - | 1 - 2 | 402 | 90 | ||
More Information
Set Membership: |
|

PIN1AT
Genomic Map