Assay Details
Gene Symbol: LARP6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129959.3 | NP_566004.1 | 9 - 10 | 1495 | 101 | |
NM_180075.2 | NP_850406.1 | 8 - 9 | 1447 | 101 | ||
GenBank mRNA | AF332565.1 | - | 9 - 10 | 1308 | 101 | |
AF367277.1 | - | 9 - 10 | 1495 | 101 | ||
AF375410.1 | - | 9 - 10 | 1434 | 101 | ||
AY056238.1 | - | 9 - 10 | 1495 | 101 | ||
AY129474.1 | - | 9 - 10 | 1308 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
