Assay Details
Gene Symbol: DL1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180087.1 | NP_850418.1 | 6 - 7 | 560 | 93 | |
NM_180088.3 | NP_850419.1 | 6 - 7 | 694 | 93 | ||
NM_180089.2 | NP_850420.1 | 7 - 8 | 753 | 93 | ||
GenBank mRNA | AF488807.1 | - | 6 - 7 | 560 | 93 | |
AJ304843.1 | - | 7 - 8 | 745 | 93 | ||
AJ304844.1 | - | 6 - 7 | 694 | 93 | ||
BT046114.1 | - | 7 - 8 | 611 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
