Assay Details
Gene Symbol: GC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180116.2 | NP_850447.1 | 11 - 12 | 1660 | 109 | |
GenBank mRNA | AY069912.1 | - | 11 - 12 | 1601 | 109 | |
BT001025.1 | - | 11 - 12 | 1511 | 109 | ||
EU249331.1 | - | 11 - 12 | 1511 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
