Assay Details
| Gene Symbol: JAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084600.1 | NP_001078069.1 | 4 - 5 | 309 | 85 | |
| NM_001202828.1 | NP_001189757.1 | 3 - 4 | 921 | 85 | ||
| NM_130200.2 | NP_566071.1 | 3 - 4 | 558 | 85 | ||
| NM_180122.4 | NP_850453.1 | 3 - 4 | 679 | 85 | ||
| GenBank mRNA | AK316746.1 | - | 3 - 4 | 478 | 85 | |
| AY150437.1 | - | 2 - 3 | 396 | 85 | ||
More Information
Set Membership: |
|

JAR1
Genomic Map