Assay Details
| Gene Symbol: JAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084600.1 | NP_001078069.1 | 5 - 6 | 1097 | 91 | |
| NM_001202828.1 | NP_001189757.1 | 4 - 5 | 1709 | 91 | ||
| NM_130200.2 | NP_566071.1 | 4 - 5 | 1346 | 91 | ||
| NM_180122.4 | NP_850453.1 | 4 - 5 | 1467 | 91 | ||
| GenBank mRNA | AK316746.1 | - | 4 - 5 | 1266 | 91 | |
| AY150437.1 | - | 3 - 4 | 1184 | 91 | ||
More Information
Set Membership: |
|

JAR1
Genomic Map