Assay Details
| Gene Symbol: SPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337245.1 | NP_001325109.1 | 4 - 5 | 1208 | 120 | |
| NM_180137.3 | NP_850468.1 | 3 - 4 | 1569 | 120 | ||
| GenBank mRNA | AJ011629.1 | - | 3 - 4 | 1122 | 120 | |
| AK230269.1 | - | 3 - 4 | 1528 | 120 | ||
More Information
Set Membership: |
|

SPL1
Genomic Map