Assay Details
Gene Symbol: SPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337245.1 | NP_001325109.1 | 6 - 7 | 1387 | 89 | |
NM_180137.3 | NP_850468.1 | 5 - 6 | 1748 | 89 | ||
GenBank mRNA | AJ011629.1 | - | 5 - 6 | 1301 | 89 | |
AK230269.1 | - | 5 - 6 | 1707 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
