Assay Details
Gene Symbol: SWI3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180145.3 | NP_850476.1 | 4 - 5 | 1255 | 126 | |
GenBank mRNA | AY062794.1 | - | 4 - 5 | 1234 | 126 | |
AY081570.1 | - | 4 - 5 | 1168 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
