Assay Details
Gene Symbol: SWI3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337268.1 | NP_001324817.1 | 5 - 6 | 1548 | 143 | |
NM_180145.3 | NP_850476.1 | 6 - 7 | 1455 | 143 | ||
GenBank mRNA | AY062794.1 | - | 6 - 7 | 1434 | 143 | |
AY081570.1 | - | 6 - 7 | 1368 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
