Assay Details
| Gene Symbol: KIN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125074.2 | NP_001118546.1 | 4 - 5 | 713 | 76 | |
| NM_110974.5 | NP_566130.1 | 4 - 5 | 829 | 76 | ||
| NM_180157.1 | NP_850488.1 | 4 - 5 | 971 | 76 | ||
| GenBank mRNA | AY093170.1 | - | 4 - 5 | 971 | 76 | |
| BT010386.1 | - | 4 - 5 | 627 | 76 | ||
| DQ778957.1 | - | 3 - 4 | 558 | 76 | ||
| X94757.1 | - | 687 | 76 | |||
More Information
Set Membership: |
|

KIN10
Genomic Map