Assay Details
Gene Symbol: SYP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111445.5 | NP_566256.1 | 5 - 6 | 1048 | 151 | |
NM_180188.3 | NP_850519.1 | 5 - 6 | 994 | 151 | ||
GenBank mRNA | AJ245408.1 | - | 5 - 6 | 970 | 151 | |
AY099723.1 | - | 5 - 6 | 994 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
