Assay Details
Gene Symbol: IMPA-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111552.3 | NP_187328.1 | 2 - 3 | 542 | 69 | |
NM_180193.3 | NP_850524.1 | 2 - 3 | 522 | 69 | ||
GenBank mRNA | AF077528.1 | - | 2 - 3 | 355 | 69 | |
AY088732.1 | - | 2 - 3 | 544 | 69 | ||
AY094390.1 | - | 2 - 3 | 368 | 69 | ||
AY125529.1 | - | 2 - 3 | 307 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
