Assay Details
| Gene Symbol: BCE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111565.6 | NP_187341.1 | 6 - 7 | 1515 | 113 | |
| NM_180196.3 | NP_850527.1 | 7 - 8 | 1430 | 113 | ||
| GenBank mRNA | AK316767.1 | - | 6 - 7 | 1413 | 113 | |
| AK317408.1 | - | 7 - 8 | 1375 | 113 | ||
| AY086441.1 | - | 6 - 7 | 1412 | 113 | ||
More Information
Set Membership: |
|

BCE2
Genomic Map