Assay Details
Gene Symbol: DWF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338420.1 | NP_001319595.1 | 2 - 3 | 1762 | 62 | |
NM_112872.3 | NP_188616.1 | 1 - 2 | 1856 | 62 | ||
NM_180285.4 | NP_850616.1 | 2 - 3 | 1353 | 62 | ||
GenBank mRNA | AB493623.1 | - | 1 - 2 | 1237 | 62 | |
AK226335.1 | - | 2 - 3 | 1290 | 62 | ||
AY072216.1 | - | 2 - 3 | 1297 | 62 | ||
AY096472.1 | - | 1 - 2 | 1237 | 62 | ||
U12400.1 | - | 1 - 2 | 1252 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
