Assay Details
| Gene Symbol: ZML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125197.2 | NP_001118669.1 | 2 - 3 | 490 | 133 | |
| NM_113012.4 | NP_566676.1 | 2 - 3 | 490 | 133 | ||
| NM_180287.5 | NP_850618.1 | 2 - 3 | 500 | 133 | ||
| GenBank mRNA | AB119060.1 | - | 2 - 3 | 372 | 133 | |
| AY042817.1 | - | 2 - 3 | 467 | 133 | ||
| AY064628.1 | - | 1 - 2 | 294 | 133 | ||
More Information
Set Membership: |
|

ZML1
Genomic Map