Assay Details
| Gene Symbol: ZML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113012.4 | NP_566676.1 | 5 - 6 | 853 | 73 | |
| NM_180287.5 | NP_850618.1 | 5 - 6 | 857 | 73 | ||
| GenBank mRNA | AB119060.1 | - | 5 - 6 | 735 | 73 | |
| AY042817.1 | - | 5 - 6 | 824 | 73 | ||
| AY064628.1 | - | 4 - 5 | 651 | 73 | ||
More Information
Set Membership: |
|

ZML1
Genomic Map