Assay Details
Gene Symbol: DCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197952.1 | NP_001184881.1 | 15 - 16 | 3988 | 108 | |
NM_099986.4 | NP_171612.1 | 15 - 16 | 4280 | 108 | ||
GenBank mRNA | AF292940.1 | - | 15 - 16 | 4255 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
