Assay Details
Gene Symbol: DCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197952.1 | NP_001184881.1 | 8 - 9 | 2429 | 70 | |
NM_099986.4 | NP_171612.1 | 8 - 9 | 2724 | 70 | ||
GenBank mRNA | AF292940.1 | - | 8 - 9 | 2699 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
