Assay Details
| Gene Symbol: CIPK9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_099996.4 | NP_171622.1 | 1 - 2 | 446 | 107 | |
| NM_179239.1 | NP_849570.1 | 1 - 2 | 297 | 107 | ||
| NM_179240.1 | NP_849571.1 | 1 - 2 | 297 | 107 | ||
| GenBank mRNA | AF339147.1 | - | 1 - 2 | 184 | 107 | |
More Information
Set Membership: |
|

CIPK9
Genomic Map