Assay Details
| Gene Symbol: AGT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342547.1 | NP_001328757.1 | 6 - 7 | 1414 | 109 | |
| NM_120126.5 | NP_568064.1 | 6 - 7 | 1359 | 109 | ||
| GenBank mRNA | AK227118.1 | - | 6 - 7 | 1221 | 109 | |
| AY054264.1 | - | 6 - 7 | 1213 | 109 | ||
| BT002306.1 | - | 6 - 7 | 1189 | 109 | ||
More Information
Set Membership: |
|

AGT2
Genomic Map