Assay Details
| Gene Symbol: HCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120135.5 | NP_568068.1 | 1 - 2 | 229 | 92 | |
| GenBank mRNA | AY087747.1 | - | 1 - 2 | 96 | 92 | |
| BT024500.1 | - | 1 - 2 | 66 | 92 | ||
More Information
Set Membership: |
|

HCC2
Genomic Map