Assay Details
| Gene Symbol: AT4G39900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120153.3 | NP_568073.1 | 3 - 4 | 468 | 122 | |
| GenBank mRNA | AY050943.1 | - | 3 - 4 | 360 | 122 | |
| AY057651.1 | - | 3 - 4 | 466 | 122 | ||
| AY079397.1 | - | 3 - 4 | 202 | 122 | ||
| AY084970.1 | - | 3 - 4 | 360 | 122 | ||
More Information
Set Membership: |
|

AT4G39900
Genomic Map