Assay Details
Gene Symbol: AT4G40050 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342566.1 | NP_001328204.1 | 3 - 4 | 1185 | 108 | |
NM_120170.3 | NP_568079.1 | 3 - 4 | 1191 | 108 | ||
GenBank mRNA | AY040014.1 | - | 3 - 4 | 1042 | 108 | |
AY133799.1 | - | 3 - 4 | 931 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
