Assay Details
| Gene Symbol: FER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120238.4 | NP_195780.1 | 4 - 5 | 998 | 120 | |
| GenBank mRNA | AF326869.1 | - | 4 - 5 | 610 | 120 | |
| AF339691.1 | - | 4 - 5 | 541 | 120 | ||
| AF412065.1 | - | 4 - 5 | 610 | 120 | ||
| AK226433.1 | - | 4 - 5 | 613 | 120 | ||
| X94248.1 | - | 579 | 120 | |||
More Information
Set Membership: |
|

FER1
Genomic Map