Assay Details
| Gene Symbol: ROPGEF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120279.3 | NP_195821.1 | 1 - 2 | 549 | 87 | |
| GenBank mRNA | BT015831.1 | - | 1 - 2 | 371 | 87 | |
| BT020213.1 | - | 1 - 2 | 209 | 87 | ||
More Information
Set Membership: |
|

ROPGEF7
Genomic Map