Assay Details
| Gene Symbol: ROPGEF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120279.3 | NP_195821.1 | 6 - 7 | 1532 | 120 | |
| GenBank mRNA | BT015831.1 | - | 6 - 7 | 1354 | 120 | |
| BT020213.1 | - | 6 - 7 | 1192 | 120 | ||
More Information
Set Membership: |
|

ROPGEF7
Genomic Map