Assay Details
| Gene Symbol: RPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120281.3 | NP_195823.1 | 2 - 3 | 1301 | 72 | |
| GenBank mRNA | AY328314.1 | - | 2 - 3 | 1234 | 72 | |
| BT020537.1 | - | 2 - 3 | 1141 | 72 | ||
More Information
Set Membership: |
|

RPL
Genomic Map