Assay Details
Gene Symbol: FHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085052.2 | NP_001078521.1 | 2 - 3 | 683 | 61 | |
NM_120298.3 | NP_195840.1 | 1 - 2 | 1324 | 61 | ||
GenBank mRNA | HM029243.1 | - | 1 - 2 | 1169 | 61 | |
HM029245.1 | - | 2 - 3 | 528 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
