Assay Details
Gene Symbol: ALG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120319.4 | NP_195861.2 | 5 - 6 | 1129 | 82 | |
GenBank mRNA | AY099540.1 | - | 5 - 6 | 1035 | 82 | |
BT002117.1 | - | 5 - 6 | 913 | 82 | ||
KJ138711.1 | - | 5 - 6 | 913 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
