Assay Details
Gene Symbol: DPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120325.4 | NP_195867.2 | 5 - 6 | 726 | 97 | |
NM_180696.2 | NP_851027.1 | 6 - 7 | 535 | 97 | ||
NM_180697.3 | NP_851028.1 | 6 - 7 | 681 | 97 | ||
GenBank mRNA | AJ294531.1 | - | 6 - 7 | 607 | 97 | |
AJ319027.1 | - | 5 - 6 | 512 | 97 | ||
AK117135.1 | - | 6 - 7 | 497 | 97 | ||
BT005286.1 | - | 5 - 6 | 434 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
