Assay Details
Gene Symbol: LCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036746.2 | NP_001031823.1 | 3 - 4 | 520 | 71 | |
NM_001342672.1 | NP_001332134.1 | 3 - 4 | 462 | 71 | ||
NM_001342673.1 | NP_001332135.1 | 3 - 4 | 462 | 71 | ||
NM_120362.4 | NP_568108.1 | 3 - 4 | 520 | 71 | ||
NM_180425.2 | NP_850756.1 | 3 - 4 | 462 | 71 | ||
GenBank mRNA | AJ937209.1 | - | 3 - 4 | 374 | 71 | |
AK317012.1 | - | 3 - 4 | 462 | 71 | ||
AY519514.1 | - | 3 - 4 | 374 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
