Assay Details
| Gene Symbol: LCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036746.2 | NP_001031823.1 | 5 - 6 | 802 | 83 | |
| NM_001342672.1 | NP_001332134.1 | 5 - 6 | 744 | 83 | ||
| NM_001342673.1 | NP_001332135.1 | 5 - 6 | 744 | 83 | ||
| NM_120362.4 | NP_568108.1 | 5 - 6 | 802 | 83 | ||
| NM_180425.2 | NP_850756.1 | 5 - 6 | 744 | 83 | ||
| GenBank mRNA | AJ937209.1 | - | 5 - 6 | 656 | 83 | |
| AK317012.1 | - | 5 - 6 | 744 | 83 | ||
| AY519514.1 | - | 5 - 6 | 656 | 83 | ||
More Information
Set Membership: |
|

LCL1
Genomic Map