Assay Details
| Gene Symbol: SBE2.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120446.4 | NP_195985.3 | 19 - 20 | 2260 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SBE2.2| Gene Symbol: SBE2.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120446.4 | NP_195985.3 | 19 - 20 | 2260 | 110 | |
Set Membership: |
|