Assay Details
| Gene Symbol: FBA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342726.1 | NP_001330597.1 | 1 - 2 | 886 | 105 | |
| NM_001342727.1 | NP_001330598.1 | 2 - 3 | 559 | 105 | ||
| NM_120450.3 | NP_568127.1 | 2 - 3 | 509 | 105 | ||
| NM_180428.2 | NP_850759.1 | 2 - 3 | 598 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001342726.1, NM_001342727.1 transcripts.
Set Membership: |
|

FBA4
Genomic Map