Assay Details
| Gene Symbol: MLS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203290.1 | NP_001190219.1 | 4 - 5 | 1075 | 117 | |
| NM_120467.4 | NP_196006.1 | 3 - 4 | 1191 | 117 | ||
| GenBank mRNA | AF360126.1 | - | 3 - 4 | 1075 | 117 | |
| AY051027.1 | - | 3 - 4 | 1032 | 117 | ||
More Information
Set Membership: |
|

MLS
Genomic Map