Assay Details
| Gene Symbol: APC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127387.3 | NP_565433.1 | 3 - 4 | 559 | 129 | |
| GenBank mRNA | AK229538.1 | - | 3 - 4 | 434 | 129 | |
| BT024912.1 | - | 3 - 4 | 326 | 129 | ||
More Information
Set Membership: |
|

APC10
Genomic Map