Assay Details
| Gene Symbol: NAP1;2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084448.2 | NP_001077917.1 | 9 - 10 | 982 | 81 | |
| NM_001202624.1 | NP_001189553.1 | 9 - 10 | 1045 | 81 | ||
| NM_127506.5 | NP_179538.1 | 9 - 10 | 982 | 81 | ||
| GenBank mRNA | AY099771.1 | - | 9 - 10 | 906 | 81 | |
| BT002602.1 | - | 9 - 10 | 837 | 81 | ||
More Information
Set Membership: |
|

NAP1;2
Genomic Map