Assay Details
Gene Symbol: FVE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127510.5 | NP_565456.2 | 1 - 2 | 532 | 70 | |
GenBank mRNA | AF498102.1 | - | 1 - 2 | 353 | 70 | |
AY057655.1 | - | 1 - 2 | 330 | 70 | ||
AY059799.1 | - | 1 - 2 | 306 | 70 | ||
AY081447.1 | - | 1 - 2 | 282 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
