Assay Details
| Gene Symbol: FVE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127510.5 | NP_565456.2 | 5 - 6 | 810 | 91 | |
| GenBank mRNA | AF498102.1 | - | 5 - 6 | 631 | 91 | |
| AY057655.1 | - | 5 - 6 | 608 | 91 | ||
| AY059799.1 | - | 5 - 6 | 584 | 91 | ||
| AY081447.1 | - | 5 - 6 | 560 | 91 | ||
More Information
Set Membership: |
|

FVE
Genomic Map